Ethyl-2-chloroacetoacetate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C1218-25G C1218-500G
TCIAC1218-25GEA 22 EUR
TCIAC1218-25G TCIAC1218-500G
Ethyl-2-chloroacetoacetate ≥95.0% (by GC)
Ethyl-2-chloroacetoacetate
Formule: C₆H₉ClO₃
Kookpunt: 77°C
Smeltpunt: –80 °C
Dichtheid: 1,19 g/cm³ (14 °C)
Vlampunt: 54°C
MDL Nummer: MFCD00009141
CAS nummer: 609-15-4
EINECS: 210-180-4
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47f1a5f120000b5bd2434010f615e4be0c7421c1&vid=47f1a5f120000b5bd2434010f615e4be0c7421c1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 % (total of isomer)