2-Chloro-6-methyl-5-nitropyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 6-Chloro-2-methyl-3-nitropyridine, 2-Chloro-5-nitro-6-picoline, 6-Chloro-3-nitro-2-picoline

C2434-1G C2434-5G
TCIAC2434-1GEA 29.7 EUR
TCIAC2434-1G TCIAC2434-5G
2-Chloro-6-methyl-5-nitropyridine ≥98.0% (by GC)
2-Chloro-6-methyl-5-nitropyridine
Formule: C₆H₅ClN₂O₂
MDL Nummer: MFCD00234160
CAS nummer: 22280-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7555409ce04146635aa4dfdbec5f605e20f9efe&vid=b7555409ce04146635aa4dfdbec5f605e20f9efe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR