2-Chlorobenzenesulphonamide 98,0 GC_ASSAY_METHOD

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: o-Chlorobenzenesulphonamide

C1990-25G C1990-500G
TCIAC1990-25GEA 44 EUR
TCIAC1990-25G TCIAC1990-500G
2-Chlorobenzenesulphonamide 98,0 GC_ASSAY_METHOD
2-Chlorobenzenesulphonamide
Formule: C₆H₆ClNO₂S
MW: 191.64 g/mol
MDL Nummer: MFCD00051974
CAS nummer: 6961-82-6
EINECS: 230-156-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc9319072e0d4d5f93cc88f07c9c4c20b997eec&vid=5fc9319072e0d4d5f93cc88f07c9c4c20b997eec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 187.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR