1-Adamantanecarbonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C0783-5G
C0783-1G
TCIAC0783-5GEA
128
EUR
TCIAC0783-5G
TCIAC0783-1G
1-Adamantanecarbonitrile ≥98.0% (by GC)
1-Adamantanecarbonitrile
Formule:
C₁₁H₁₅N Kookpunt: 275 °C (1013 hPa) Smeltpunt: 185…187 °C Dichtheid: 1,13 g/cm³ (25 °C) |
MDL Nummer:
MFCD00074731 CAS nummer: 23074-42-2 EINECS: 245-413-9 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...