Ethyl-2-methylacetoacetate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ethyl 2-methyl-3-oxobutanoate
M0698-25ML M0698-250ML
TCIAM0698-25MLEA 27.5 EUR
TCIAM0698-25ML TCIAM0698-250ML
Ethyl-2-methylacetoacetate ≥95.0% (by GC)
Ethyl-2-methylacetoacetate
Formule: C₇H₁₂O₃
MW: 144.17 g/mol
Kookpunt: 185°C
Smeltpunt: -45
Vlampunt: 62°C
MDL Nummer: MFCD00009164
CAS nummer: 609-14-3
EINECS: 210-179-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cf29a8afd158a9ac361ba30251bb982dc408be8&vid=9cf29a8afd158a9ac361ba30251bb982dc408be8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %