2-Hydroxyacetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H0347-25G
H0347-5G
TCIAH0347-25GEA
115
EUR
TCIAH0347-25G
TCIAH0347-5G
2-Hydroxyacetophenone ≥98.0% (by GC)
2-Hydroxyacetophenone
Formule:
C₈H₈O₂ Kookpunt: 126°C Smeltpunt: 103…105 °C Dichtheid: 1,0963 g/cm³ (99 °C) |
MDL Nummer:
MFCD00041829 CAS nummer: 582-24-1 EINECS: 209-480-8 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 85.0 to 88.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...