Methylprednisolone ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M1665-1G M1665-5G
TCIAM1665-1GEA 78.1 EUR
TCIAM1665-1G TCIAM1665-5G
Methylprednisolone ≥98.0% (by HPLC)
Methylprednisolone
Formule: C₂₂H₃₀O₅
Smeltpunt: 237°C
MDL Nummer: MFCD00010591
CAS nummer: 83-43-2
EINECS: 201-476-4
Merck Index: 14,06111
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a439fb1e497b8828a8da9848f89c40c1ec6ed76&vid=6a439fb1e497b8828a8da9848f89c40c1ec6ed76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +79.0 to +86.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR