2-Ethoxybenzamide ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E0222-25G
E0222-500G
TCIAE0222-25GEA
18.7
EUR
TCIAE0222-25G
TCIAE0222-500G
2-Ethoxybenzamide ≥98.0% (by GC)
2-Ethoxybenzamide
Formule:
C₉H₁₁NO₂ Kookpunt: 333 °C (1013 hPa) Smeltpunt: 132…134 °C |
MDL Nummer:
MFCD00007977 CAS nummer: 938-73-8 EINECS: 213-346-4 Merck Index: 14,03731 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...