(4-Aminophenyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1289-25G A1289-5G
TCIAA1289-25GEA 648 EUR
TCIAA1289-25G TCIAA1289-5G
(4-Aminophenyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)
(4-Aminophenyl)acetic acid ethyl ester
Formule: C₁₀H₁₃NO₂
MDL Nummer: MFCD00017569
CAS nummer: 5438-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c3e90e9772965c3458a484ee9e59d3a25a58f03&vid=3c3e90e9772965c3458a484ee9e59d3a25a58f03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR