Ethyl nitroacetate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
N0657-25G N0657-5G
TCIAN0657-25GEA 91.3 EUR
TCIAN0657-25G TCIAN0657-5G
Ethyl nitroacetate ≥95.0% (by GC)
Ethyl nitroacetate
Formule: C₄H₇NO₄
Kookpunt: 107°C
Dichtheid: 1,226 g/cm³ (0 °C)
Vlampunt: 92°C
MDL Nummer: MFCD00007403
CAS nummer: 626-35-7
EINECS: 210-944-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ffe933382ea8afbd9182888760fbbba54fdb8b&vid=74ffe933382ea8afbd9182888760fbbba54fdb8b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %