(4-Methoxyphenyl)acetic acid ethyl ester ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0793-25ML M0793-500ML
TCIAM0793-25MLEA 64.9 EUR
TCIAM0793-25ML TCIAM0793-500ML
(4-Methoxyphenyl)acetic acid ethyl ester ≥98.0% (by GC)
(4-Methoxyphenyl)acetic acid ethyl ester
Formule: C₁₁H₁₄O₃
Kookpunt: 158°C
Dichtheid: 1,097 g/cm³ (24 °C)
MDL Nummer: MFCD00040760
CAS nummer: 14062-18-1
EINECS: 237-903-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73f4491e7154bee648df0718d657ed684b111f51&vid=73f4491e7154bee648df0718d657ed684b111f51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR