Ethyl diethoxyacetate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D1883-25ML
D1883-500ML
D1883-100ML
TCIAD1883-25MLEA
55
EUR
TCIAD1883-25ML
TCIAD1883-500ML
TCIAD1883-100ML
Ethyl diethoxyacetate ≥97.0% (by GC)
Ethyl diethoxyacetate
Formule:
C₈H₁₆O₄ Kookpunt: 84°C Dichtheid: 0,994 g/cm³ (18 °C) Vlampunt: 85°C |
MDL Nummer:
MFCD00009231 CAS nummer: 6065-82-3 EINECS: 227-999-8 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...