Methoxyazijnzuur ≥96.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
M0827-25G
M0827-500G
TCIAM0827-25GEA
17.6
EUR
TCIAM0827-25G
TCIAM0827-500G
Methoxyazijnzuur ≥96.0% (by titrimetric analysis)
Methoxyazijnzuur
Formule:
C₃H₆O₃ Kookpunt: 204°C Smeltpunt: 7 °C Dichtheid: 1,18 g/cm³ (20 °C) Vlampunt: 125°C |
MDL Nummer:
MFCD00004308 CAS nummer: 625-45-6 EINECS: 210-894-6 UN: 3265 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...