2-(Diphenylphosphino)benzoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D3242-1G
D3242-5G
TCIAD3242-1GEA
36.3
EUR
TCIAD3242-1G
TCIAD3242-5G
2-(Diphenylphosphino)benzoic acid ≥98.0% (by GC, titration analysis)
2-(Diphenylphosphino)benzoic acid
Formule:
C₁₉H₁₅O₂P MW: 306.3 g/mol |
MDL Nummer:
MFCD00674024 CAS nummer: 17261-28-8 EINECS: 241-293-7 |
|
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 177.0 to 181.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...