Diphenyl carbonate ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C0043-25G C0043-500G
TCIAC0043-25GEA 18.7 EUR
TCIAC0043-25G TCIAC0043-500G
Diphenyl carbonate ≥99.0% (by GC)
Diphenyl carbonate
Formule: C₁₃H₁₀O₃
Kookpunt: 301 °C (1013 hPa)
Smeltpunt: 78 °C
Dichtheid: 1,272 g/cm³ (14 °C)
Vlampunt: 168 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003037
CAS nummer: 102-09-0
EINECS: 203-005-8
UN: 3077
ADR: 9,III
Merck Index: 14,07278
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1cb0d00429f49103d09ec60adf65b815c427a8f&vid=d1cb0d00429f49103d09ec60adf65b815c427a8f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White powder to crystal
Purity (GC) min. 99.0 %
Melting point 79.0 to 82.0 deg-C
Solubility in Methanol almost transparency