Thiobenzoic acid ≥93.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T0195-25G T0195-500G
TCIAT0195-25GEA 17.6 EUR
TCIAT0195-25G TCIAT0195-500G
Thiobenzoic acid ≥93.0% (by titrimetric analysis)
Thiobenzoic acid
Formule: C₇H₆OS
Kookpunt: 87°C
Smeltpunt: 24 °C
Dichtheid: 1,1775 g/cm³ (20 °C)
Vlampunt: 94°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004852
CAS nummer: 98-91-9
EINECS: 202-712-9
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37921c4a42e7e02540ce1a204376c2a365d8c199&vid=37921c4a42e7e02540ce1a204376c2a365d8c199&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to lump to clear liquid
Purity (Iodometric Titration) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR