3-Methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Methyl-5-(trifluoromethyl)pyrazole
M2032-1G
M2032-5G
TCIAM2032-1GEA
18.7
EUR
TCIAM2032-1G
TCIAM2032-5G
3-Methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
3-Methyl-5-(trifluoromethyl)-1H-pyrazole
Formule:
C₅H₅F₃N₂ |
MDL Nummer:
MFCD00156058 CAS nummer: 10010-93-2 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...