2-Fluoro-5-methoxybenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
F0642-1G
F0642-5G
TCIAF0642-1GEA
24.2
EUR
TCIAF0642-1G
TCIAF0642-5G
2-Fluoro-5-methoxybenzonitrile ≥98.0% (by GC)
2-Fluoro-5-methoxybenzonitrile
Formule:
C₈H₆FNO |
MDL Nummer:
MFCD00142921 CAS nummer: 127667-01-0 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...