Ethyl-4,4,4-trifluoroacetoacetate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate
T0810-25G T0810-500G T0810-100G
TCIAT0810-25GEA 51.7 EUR
TCIAT0810-25G TCIAT0810-500G TCIAT0810-100G
Ethyl-4,4,4-trifluoroacetoacetate ≥98.0% (by GC)
Ethyl-4,4,4-trifluoroacetoacetate
Formule: C₆H₇F₃O₃
MW: 184.11 g/mol
Kookpunt: 132°C
Smeltpunt: -39
Vlampunt: 39°C
MDL Nummer: MFCD00000424
CAS nummer: 372-31-6
EINECS: 206-750-7
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f32e2d401c6336dd3edd52e7eb1586a63c1b292&vid=7f32e2d401c6336dd3edd52e7eb1586a63c1b292&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %