Methyl benzenesulphonate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0033-25ML
B0033-500ML
B0033-100ML
TCIAB0033-25MLEA
24.2
EUR
TCIAB0033-25ML
TCIAB0033-500ML
TCIAB0033-100ML
Methyl benzenesulphonate ≥98.0% (by GC)
Methyl benzenesulphonate
Formule:
C₇H₈O₃S Kookpunt: 280°C Dichtheid: 1,2889 g/cm³ (0 °C) Vlampunt: 143°C |
MDL Nummer:
MFCD00014737 CAS nummer: 80-18-2 EINECS: 201-256-8 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...