Coronene ≥80.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C0386-1G C0386-5G
TCIAC0386-1GEA 114 EUR
TCIAC0386-1G TCIAC0386-5G
Coronene ≥80.0% (by HPLC)
Coronene
Formule: C₂₄H₁₂
Kookpunt: 525°C
Smeltpunt: 440°C
Dichtheid: 1,371 g/cm³ (25 °C)
MDL Nummer: MFCD00004134
CAS nummer: 191-07-1
EINECS: 205-881-7
Merck Index: 14,02536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a56f09eb6369d4f43d33169d6ca657637a64c42c&vid=a56f09eb6369d4f43d33169d6ca657637a64c42c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 80.0 area%