Cyclohexyl acetate ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A0028-25ML A0028-500ML
TCIAA0028-25MLEA 17.6 EUR
TCIAA0028-25ML TCIAA0028-500ML
Cyclohexyl acetate ≥99.0% (by GC)
Cyclohexyl acetate
Formule: C₈H₁₄O₂
Kookpunt: 174°C
Smeltpunt: –77 °C
Dichtheid: 0,97 g/cm³ (20 °C)
Vlampunt: 58°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003850
CAS nummer: 622-45-7
EINECS: 210-736-6
UN: 2243
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eec09b36919476a6e02aede149328fd7733b9a7d&vid=6294f6b4a0d356d3e8966aac3802e030d5d8339a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %