Arachidic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid

E0006-1G E0006-25G
TCIAE0006-1GEA 17.6 EUR
TCIAE0006-1G TCIAE0006-25G
Arachidic acid ≥98.0% (by GC, titration analysis)
Arachidic acid
Formule: C₂₀H₄₀O₂
Kookpunt: 303°C
Dichtheid: (literatuur)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00002755
CAS nummer: 506-30-9
EINECS: 208-031-3
Merck Index: 14,00764
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d33bd761fc1b0388163d10611a9598ce58d40dc&vid=9e6c92b69ad734eeae45abe77f1c749146f76d52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 75.0 to 78.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR