Pentacosanoic acid ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P0882-1G
P0882-5G
TCIAP0882-1GEA
198
EUR
TCIAP0882-1G
TCIAP0882-5G
Pentacosanoic acid ≥96.0% (by GC)
Pentacosanoic acid
Formule:
C₂₅H₅₀O₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00020551 CAS nummer: 506-38-7 EINECS: 208-036-0 |
|
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 96.0 % |
Melting point | 82.0 to 85.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...