Perchloroethane ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H0060-25G
H0060-500G
H0060-100G
TCIAH0060-25GEA
17.6
EUR
TCIAH0060-25G
TCIAH0060-500G
TCIAH0060-100G
Perchloroethane ≥99.0% (by GC)
Perchloroethane
Formule:
C₂Cl₆ Kookpunt: 189 °C (1013 hPa) Dichtheid: 2,091 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000799 CAS nummer: 67-72-1 EINECS: 200-666-4 UN: 3077 ADR: 9,III Merck Index: 14,04679 |
|
Specification Test Results
Appearance | White powder to crystal |
Purity (GC) | min. 99.0 % |
Solubility in Methanol | faint turbidity |
Melting point | 185.0 to 189.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...