1,3-Dichloroacetone ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D0401-25G
D0401-500G
TCIAD0401-25GEA
36.3
EUR
TCIAD0401-25G
TCIAD0401-500G
1,3-Dichloroacetone ≥98.0% (by GC)
1,3-Dichloroacetone
Formule:
ClH₂CC(O)CH₂Cl Kookpunt: 173°C Smeltpunt: 39…42 °C Dichtheid: 1,432 g/cm³ (20 °C) Vlampunt: 95 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00000937 CAS nummer: 534-07-6 EINECS: 208-585-6 UN: 2649 ADR: 6.1,II Merck Index: 14,03052 |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...