2,2,2-Trichloroethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2,2,2-Trichloroethan-1-ol

T0622-25G T0622-500G T0622-100G
TCIAT0622-25GEA 24.2 EUR
TCIAT0622-25G TCIAT0622-500G TCIAT0622-100G
2,2,2-Trichloroethanol ≥98.0% (by GC)
2,2,2-Trichloroethanol
Formule: C₂H₃Cl₃O
Kookpunt: 151°C
Smeltpunt: 17…18 °C
Dichtheid: 1,56 g/cm³ (20 °C)
Vlampunt: ≥ 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004677
CAS nummer: 115-20-8
EINECS: 204-071-0
UN: 1760
ADR: 8,III
Merck Index: 14,09638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c471b7dd8799d4686d80944ba0fc9f6e47555331&vid=e560074141569b4b4c6cfeb0abf5a87537349a2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR