3-Chloor-1-propanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Chloropropan-1-ol, 1-Chloro-3-hydroxypropane
C0270-25G
C0270-500G
C0270-100G
TCIAC0270-25GEA
37.4
EUR
TCIAC0270-25G
TCIAC0270-500G
TCIAC0270-100G
3-Chloor-1-propanol ≥98.0% (by GC)
3-Chloor-1-propanol
Formule:
C₃H₇ClO MW: 94.54 g/mol Kookpunt: 160°C Smeltpunt: -20 Vlampunt: 82°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002943 CAS nummer: 627-30-5 EINECS: 210-992-9 UN: 2849 ADR: 6.1 III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...