1,1,1-Trifluoroiodoethane ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Iodo-2,2,2-trifluoroethane, 2,2,2-Trifluoroethyliodide, 2-Iodo-1,1,1-trifluoroethane, 2,2,2-Trifluoroiodoethane, 1,1,1-Trifluoro-2-iodoethane
T1148-25G T1148-5G
TCIAT1148-25GEA 170 EUR
TCIAT1148-25G TCIAT1148-5G
1,1,1-Trifluoroiodoethane ≥99.0% (by GC)
1,1,1-Trifluoroiodoethane
Formule: C₂H₂F₃I
Kookpunt: 55°C
Dichtheid: 2,1499 g/cm³ (20 °C)
MDL Nummer: MFCD00001082
CAS nummer: 353-83-3
EINECS: 206-541-0
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01f6412c3b64de1f592bfb68895a18b38e35dcb3&vid=8a6a74c3abcfeed85493a0aeecd3027cd3736e19&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %