2,2,3,3-Tetrafluoro-1-propanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,2,3,3-Tetrafluoropropan-1-ol, 1H,1H,3H-Tetrafluoro-1-propanol

T0101-25G T0101-500G T0101-100G
TCIAT0101-25GEA 23.1 EUR
TCIAT0101-25G TCIAT0101-500G TCIAT0101-100G
2,2,3,3-Tetrafluoro-1-propanol ≥98.0% (by GC)
2,2,3,3-Tetrafluoro-1-propanol
Formule: C₃H₄F₄O
Kookpunt: 106°C
Smeltpunt: –15 °C
Dichtheid: 1,6647 g/cm³ (20 °C)
Vlampunt: 45°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004676
CAS nummer: 76-37-9
EINECS: 200-955-5
UN: 1993
ADR: 3,III
Merck Index: 14,09206
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dc51da2ec27680ef867d2f805c6608627dc32e8&vid=170e2f6b639236a9a5e63236b949d7983e39be34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR