Tributylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0357-25ML T0357-500ML
TCIAT0357-25MLEA 18.7 EUR
TCIAT0357-25ML TCIAT0357-500ML
Tributylamine ≥98.0% (by GC, titration analysis)
Tributylamine
Formule: C₁₂H₂₇N
MW: 185.35 g/mol
Kookpunt: 217°C
Smeltpunt: –70°C
Vlampunt: 74°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009431
CAS nummer: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1 II
Merck Index: 14,09618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=241763504005ba63b2472e70d918896745887279&vid=6de81f60e9e5d4502d36b2033877dac4f5ffe8ba&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %