Trihexylamine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Tri-n-hexylamine, Tri(hex-1-yl)amine

T0442-25ML T0442-500ML
TCIAT0442-25MLEA 49.5 EUR
TCIAT0442-25ML TCIAT0442-500ML
Trihexylamine ≥95.0% (by GC)
Trihexylamine
Formule: C₁₈H₃₉N
MW: 269.51 g/mol
Kookpunt: 265°C
Smeltpunt: -75
Vlampunt: 132°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009523
CAS nummer: 102-86-3
EINECS: 203-062-9
UN: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af369cb078e266cb8ed6cfea8e3cca7faa5580c0&vid=c41ae93bf58a4f66829128cab00f47ade884abfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR