Butylamine ≥99.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Butylamine, 1-Aminobutane, n-Butylamine
B0707-25ML
B0707-500ML
TCIAB0707-25MLEA
17.6
EUR
TCIAB0707-25ML
TCIAB0707-500ML
Butylamine ≥99.0% (by GC, titration analysis)
Butylamine
Formule:
H₃C(CH₂)₂CH₂NH₂ MW: 73.14 g/mol Kookpunt: 77°C Smeltpunt: -50 Vlampunt: –4°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011690 CAS nummer: 109-73-9 EINECS: 203-699-2 UN: 1125 ADR: 3 II Merck Index: 14,01543 |
|
Specification Test Results
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...