Pentadecylamine ≥95.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0763-10G A0763-5G
TCIAA0763-10GEA 318 EUR
TCIAA0763-10G TCIAA0763-5G
Pentadecylamine ≥95.0% (by GC, titration analysis)
Pentadecylamine
Formule: H₃C(CH₂)₁₃CH₂NH₂
Kookpunt: 307 °C (1013 hPa)
Smeltpunt: 33…35 °C
Dichtheid: 0,9119 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00038195
CAS nummer: 2570-26-5
EINECS: 219-912-7
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fd5f3670e3cfed1eb5c4658519d8bde8d391eb4&vid=7fd5f3670e3cfed1eb5c4658519d8bde8d391eb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 35.0 to 39.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR