Tetradecylamine ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0090-25G T0090-400G
TCIAT0090-25GEA 18.7 EUR
TCIAT0090-25G TCIAT0090-400G
Tetradecylamine ≥96.0% (by GC)
Tetradecylamine
Formule: H₃C(CH₂)₁₂CH₂NH₂
Kookpunt: 291°C
Smeltpunt: 40 °C
Dichtheid: 0,8571 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008157
CAS nummer: 2016-42-4
EINECS: 217-950-9
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e4013a4ac46ad02f5052e5f15b256011fa6e4c5&vid=fe3a98ac7b1b9de7cc300f02b005fe35ef8ffdf4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR