Isopropylamine ≥99.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Aminopropane
I0165-25ML
I0165-500ML
TCIAI0165-25MLEA
17.6
EUR
TCIAI0165-25ML
TCIAI0165-500ML
Isopropylamine ≥99.0% (by GC, titration analysis)
Isopropylamine
Formule:
C₃H₉N Kookpunt: 34°C Smeltpunt: –95°C Dichtheid: 0,69 g/cm³ (20 °C) Vlampunt: –20°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008082 CAS nummer: 75-31-0 EINECS: 200-860-9 UN: 1221 ADR: 3,I Merck Index: 14,05209 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...