Formaldehyde dimethyl acetal ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Formal, Methylal, Dimethoxymethane

D0637-25ML D0637-500ML
TCIAD0637-25MLEA 17.6 EUR
TCIAD0637-25ML TCIAD0637-500ML
Formaldehyde dimethyl acetal ≥98.0% (by GC)
Formaldehyde dimethyl acetal
Formule: CH₂(OCH₃)₂
MW: 76.1 g/mol
Kookpunt: 42°C
Smeltpunt: -105
Vlampunt: –32°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008495
CAS nummer: 109-87-5
EINECS: 203-714-2
UN: 1234
ADR: 3 II
Merck Index: 14,06012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b29644311497472fab8d7152520936a239dedc&vid=ad6b8d30fdf42b49b1326bd5f49cfda3fd15d9b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR