1-Hexacosanol ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H0342-1G H0342-100MG
TCIAH0342-1GEA 388 EUR
TCIAH0342-1G TCIAH0342-100MG
1-Hexacosanol ≥95.0% (by GC)
1-Hexacosanol
Formule: H₃C(CH₂)₂₄CH₂OH
Kookpunt: 412 °C (1013 hPa)
Smeltpunt: 77…78 °C
Dichtheid: 0,8215 g/cm³ (85 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002940
CAS nummer: 506-52-5
EINECS: 208-044-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a0f95437c5a5a40980cbf5156c242146fe29229&vid=2bf521caad1e9abfd34b388ebe58aaa3fc16661a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 78.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR