1-Nonadecanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

N0284-25G N0284-5G
TCIAN0284-25GEA 293 EUR
TCIAN0284-25G TCIAN0284-5G
1-Nonadecanol ≥98.0% (by GC)
1-Nonadecanol
Formule: H₃C(CH₂)₁₇CH₂OH
Kookpunt: 345 °C (1013 hPa)
Smeltpunt: 60…61 °C
Dichtheid: 0,882 g/cm³ (21 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002824
CAS nummer: 1454-84-8
EINECS: 215-930-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f1e6f95c175934fdcb4a90f55aad1fc71a2e8cb&vid=373d830fa5608fb0469d0a943564bf6c0b0787eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 65.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR