1-Pentadecanol ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P0036-25G
P0036-500G
P0036-100G
TCIAP0036-25GEA
86.9
EUR
TCIAP0036-25G
TCIAP0036-500G
TCIAP0036-100G
1-Pentadecanol ≥95.0% (by GC)
1-Pentadecanol
Formule:
H₃C(CH₂)₁₃CH₂OH Kookpunt: 133°C Smeltpunt: 41…44 °C Dichtheid: 0,893 g/cm³ (21 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004759 CAS nummer: 629-76-5 EINECS: 211-107-9 |
|
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 95.0 % |
Melting point | 42.0 to 46.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...