1-Tetracosanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T0593-1G
T0593-5G
TCIAT0593-1GEA
104
EUR
TCIAT0593-1G
TCIAT0593-5G
1-Tetracosanol ≥98.0% (by GC)
1-Tetracosanol
Formule:
H₃C(CH₂)₂₂CH₂OH Kookpunt: 190 °C (1013 hPa) Smeltpunt: 81…83 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00042662 CAS nummer: 506-51-4 EINECS: 208-043-9 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 77.0 deg-C |
Solubility in hot EtOH | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...