(±)-2-Decanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (±)-Methyl octyl carbinol, (±)-2-Hydroxy decane, 2-Decanol, (±)-Octyl methyl carbinol
D1379-25ML D1379-5ML
TCIAD1379-25MLEA 323 EUR
TCIAD1379-25ML TCIAD1379-5ML
(±)-2-Decanol ≥98.0% (by GC)
(±)-2-Decanol
Formule: C₁₀H₂₂O
Kookpunt: 211°C
Smeltpunt: –5 °C
Dichtheid: 0,827 g/cm³ (20 °C)
Vlampunt: 80°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004594
CAS nummer: 1120-06-5
EINECS: 214-296-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24fc5195d7f1b8a9f21d7d40fe97096f1d6d1914&vid=3f2d4211427d52537718c968cb5d33940a66de6a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %