(±)-2-Undecanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Undecan-2-ol, (±)-Methyl nonyl carbinol, (±)-2-Hydroxy undecane, (±)-Nonyl methyl carbinol, 2-Undecanol
U0027-25ML U0027-5ML
TCIAU0027-25MLEA 57.2 EUR
TCIAU0027-25ML TCIAU0027-5ML
(±)-2-Undecanol ≥98.0% (by GC)
(±)-2-Undecanol
Formule: C₁₁H₂₄O
MW: 172.31 g/mol
Kookpunt: 228°C
Vlampunt: 113°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00021958
CAS nummer: 1653-30-1
EINECS: 216-722-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37620d438f4a09140b492f5fd7ee167d654e0a25&vid=dab976921e9631af4e6a5f316d00e9eb8e458459&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %