3-Pentanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P0057-25ML
P0057-500ML
TCIAP0057-25MLEA
17.6
EUR
TCIAP0057-25ML
TCIAP0057-500ML
3-Pentanol ≥98.0% (by GC)
3-Pentanol
Formule:
C₅H₁₂O Kookpunt: 116°C Smeltpunt: –8°C Dichtheid: 0,82 g/cm³ (20 °C) Vlampunt: 34°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004574 CAS nummer: 584-02-1 EINECS: 209-526-7 UN: 1105 ADR: 3,III Merck Index: 14,07120 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...