1-Heptanethiol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: n-Heptanethiol, n-Heptyl mercaptan, 1-Mercaptoheptane

H0029-10G H0029-25G
TCIAH0029-10GEA 51.7 EUR
TCIAH0029-10G TCIAH0029-25G
1-Heptanethiol ≥98.0% (by GC)
1-Heptanethiol
Formule: C₇H₁₆S
Kookpunt: 177°C
Smeltpunt: -43°C
Dichtheid: 0,854 g/cm³ (20 °C)
Vlampunt: 46°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004911
CAS nummer: 1639-09-4
EINECS: 216-678-8
UN: 3336
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5267787e778bc7c9bdc700d8c0db05f8e6564708&vid=0e368eeb87125930457ec13c5a7a5a6b0d9c7322&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR