1-Hexadecanethiol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H0068-25ML H0068-500ML
TCIAH0068-25MLEA 17.6 EUR
TCIAH0068-25ML TCIAH0068-500ML
1-Hexadecanethiol ≥97.0% (by GC)
1-Hexadecanethiol
Formule: C₁₆H₃₄S
Kookpunt: 189°C
Smeltpunt: 18…20 °C
Dichtheid: 0,8556 g/cm³ (20 °C)
Vlampunt: 135°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011677
CAS nummer: 2917-26-2
EINECS: 220-846-6
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ca9c63c8de4f3b0f0d5eba82de60b8c6ba79f7b&vid=3c9dc6e9b3b84d2a27a6fd1146e4bf8cb7a94254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR