tert-Butyl propiolate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P1038-25G P1038-5G
TCIAP1038-25GEA 428 EUR
TCIAP1038-25G TCIAP1038-5G
tert-Butyl propiolate ≥98.0% (by GC)
tert-Butyl propiolate
Formule: C₇H₁₀O₂
Kookpunt: 53°C
Vlampunt: 28°C
MDL Nummer: MFCD00060100
CAS nummer: 13831-03-3
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32c292c38d3b71a8c799ed2213c4522adb11a9d0&vid=32c292c38d3b71a8c799ed2213c4522adb11a9d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR