1-(Trimethylsilyl)-1-propyne ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Trimethyl-1-propynylsilane

T1123-100ML T1123-25ML T1123-5ML
TCIAT1123-100MLEA 273 EUR
TCIAT1123-100ML TCIAT1123-25ML TCIAT1123-5ML
1-(Trimethylsilyl)-1-propyne ≥98.0% (by GC)
1-(Trimethylsilyl)-1-propyne
Formule: C₆H₁₂Si
MW: 112.25 g/mol
Kookpunt: 100°C
Dichtheid: 0.759
Vlampunt: –4°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009271
CAS nummer: 6224-91-5
EINECS: 228-314-5
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e68fc5abc6fa38b17cb3ee204d930a8ee8069e7&vid=6e68fc5abc6fa38b17cb3ee204d930a8ee8069e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR