Potassium cyclopropyltrifluoroborate ≥95.0% (W)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Cyclopropyltrifluoroboric acid potassium salt

P1846-1G P1846-5G
TCIAP1846-1GEA 40.7 EUR
TCIAP1846-1G TCIAP1846-5G
Potassium cyclopropyltrifluoroborate ≥95.0% (W)
Potassium cyclopropyltrifluoroborate
Formule: C₃H₅BF₃K
MW: 147.98 g/mol
MDL Nummer: MFCD09265031
CAS nummer: 1065010-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0fe5148068e20df021639fd459fd5112a2cb82a&vid=e0fe5148068e20df021639fd459fd5112a2cb82a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (with ignition residue) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR