1-Bromo-2-methylpropane ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Isobutyl bromide
B0616-25G
B0616-500G
TCIAB0616-25GEA
17.6
EUR
TCIAB0616-25G
TCIAB0616-500G
1-Bromo-2-methylpropane ≥97.0% (by GC)
1-Bromo-2-methylpropane
Formule:
C₄H₉Br MW: 137.02 g/mol Kookpunt: 92°C Smeltpunt: -118 Vlampunt: 18°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000217 CAS nummer: 78-77-3 EINECS: 201-141-2 UN: 2342 ADR: 3 II Merck Index: 13,05152 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...